Potassium (Fluorosulphonyl)(trifluoromethanesulphonyl)imide ≥97.0% (by titrimetric analysis)
Proveedor: TCI
|
Peligro
|
P2410-1G
P2410-5G
TCIAP2410-1GEA
97
EUR
TCIAP2410-1G
TCIAP2410-5G
Potassium (Fluorosulphonyl)(trifluoromethanesulphonyl)imide ≥97.0% (by titrimetric analysis)
Potassium (Fluorosulphonyl)(trifluoromethanesulphonyl)imide
Formula:
CF₄KNO₄S₂ MW: 269,23 g/mol Pto. de fusión: 101°C |
Número MDL:
MFCD28334505 Núm. CAS: 860653-59-4 UN: 1759 ADR: 8,II |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (Ion exchange titration) | min. 97.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...