5alpha-Pregnane-3,20-dione ≥98.0% (por GC)
Proveedor: TCI
Sinónimos:
3,20-Allopregnanedione, 5alpha-Dihydroprogesterone
P2423-1G
P2423-5G
TCIAP2423-1GEA
63
EUR
TCIAP2423-1G
TCIAP2423-5G
5alpha-Pregnane-3,20-dione ≥98.0% (por GC)
5alpha-Pregnane-3,20-dione
Formula:
C₂₁H₃₂O₂ MW: 316,49 g/mol |
Número MDL:
MFCD00067132 Núm. CAS: 566-65-4 EINECS: 209-297-3 Índice Merck: 14,00272 |
Resultados test especificaciones
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 202.0 to 206.0 deg-C |
Specific rotation [a]20/D | 113 to 123 deg (C=1, CHCL3) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...