trans-4-(trans-4-Propylcyclohexyl)cyclohexanol ≥98.0% (por GC)
Proveedor: TCI
P2434-1G
P2434-5G
TCIAP2434-1GEA
31
EUR
TCIAP2434-1G
TCIAP2434-5G
trans-4-(trans-4-Propylcyclohexyl)cyclohexanol ≥98.0% (por GC)
trans-4-(trans-4-Propylcyclohexyl)cyclohexanol
Formula:
C₁₅H₂₈O |
Número MDL:
MFCD09752068 Núm. CAS: 82832-72-2 |
Resultados test especificaciones
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 122.0 to 126.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...