Posaconazole ≥98.0% (by HPLC)
Proveedor: TCI
|
Atención
|
P2477-10MG
P2477-50MG
TCIAP2477-10MGEA
32
EUR
TCIAP2477-10MG
TCIAP2477-50MG
Posaconazole ≥98.0% (by HPLC)
Posaconazole
Formula:
C₃₇H₄₂F₂N₈O₄ MW: 700,79 g/mol Temperatura de almacenaje: Congelador |
Número MDL:
MFCD00941162 Núm. CAS: 171228-49-2 Índice Merck: 14,07602 |
Resultados test especificaciones
Appearance | White to Light yellow powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Elemental analysis (Nitrogen) | 15.00 to 17.00 % |
Melting point | 167.0 to 171.0 deg-C |
Specific rotation [a]20 | -24.0 to -28.0 deg (C=1, CHCL3) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...