Pyrrolidine Hydriodide ≥98.0% (by total nitrogen and titration analysis)

Proveedor: TCI
Atención

Sinónimos: Pyrrolidinium iodide

P2486-1G P2486-5G
TCIAP2486-1GEA 42 EUR
TCIAP2486-1G TCIAP2486-5G
Pyrrolidine Hydriodide ≥98.0% (by total nitrogen and titration analysis)
Pyrrolidine Hydriodide
Formula: C₄H₉N·HI
MW: 199,04 g/mol
Núm. CAS: 45361-12-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ccd5fb358ea0e02428a688c5659b007d665e3923&vid=ccd5fb358ea0e02428a688c5659b007d665e3923&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (with Total Nitrogen) min. 98.0 %
Purity (Argentmetric Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR