Piperazine dihydrobromide ≥98.0% (by total nitrogen and titration analysis)
Proveedor: TCI
|
Atención
|
Sinónimos:
Piperazinium Dibromide, Piperazinium bromide
P2490-1G
P2490-5G
TCIAP2490-1GEA
30
EUR
TCIAP2490-1G
TCIAP2490-5G
Piperazine dihydrobromide ≥98.0% (by total nitrogen and titration analysis)
Piperazine dihydrobromide
Formula:
C₄H₁₀N₂·₂HBr MW: 247,96 g/mol Pto. de fusión: 360°C |
Núm. CAS:
59813-05-7 EINECS: 261-937-0 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (with Total Nitrogen) | min. 98.0 % |
Purity (Argentmetric Titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...