Piperazine dihydriodide ≥98.0% (by total nitrogen and titration analysis)

Proveedor: TCI
Atención

Sinónimos: Piperazinium Diiodide, Piperazinium iodide

P2492-1G P2492-5G
TCIAP2492-1GEA 56 EUR
TCIAP2492-1G TCIAP2492-5G
Piperazine dihydriodide ≥98.0% (by total nitrogen and titration analysis)
Piperazine dihydriodide
Formula: C₄H₁₀N₂·₂HI
MW: 341.96 g/mol
Pto. de fusión: 250°C
Núm. CAS: 58464-47-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f97868beac6b50f462fe8a3210e66667c31d25c0&vid=f97868beac6b50f462fe8a3210e66667c31d25c0&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Light yellow to Light orange powder to crystal
Purity (with Total Nitrogen) min. 98.0 %
Purity (Argentmetric Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR