2-(9,9'-Spirobi[fluoren]-2-yl)-4,6-diphenyl-1,3,5-triazine ≥98.0% (by HPLC)

Proveedor: TCI

Sinónimos: 2,4-Diphenyl-6-(9,9'-spirobi[9H-fluoren]-2-yl)-1,3,5-triazine

P2512-1G P2512-200MG
TCIAP2512-1GEA 209 EUR
TCIAP2512-1G TCIAP2512-200MG
2-(9,9'-Spirobi[fluoren]-2-yl)-4,6-diphenyl-1,3,5-triazine ≥98.0% (by HPLC)
2-(9,9'-Spirobi[fluoren]-2-yl)-4,6-diphenyl-1,3,5-triazine
Formula: C₄₀H₂₅N₃
MW: 547,66 g/mol
Número MDL: MFCD30721570
Núm. CAS: 1207176-84-8

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR