pentaceno, purified by sublimation
Proveedor: TCI
P2524-100MG
P2524-1G
TCIAP2524-100MGEA
31
EUR
TCIAP2524-100MG
TCIAP2524-1G
pentaceno, purified by sublimation
pentaceno
Formula:
C₂₂H₁₄ |
Número MDL:
MFCD00003710 Núm. CAS: 135-48-8 EINECS: 205-193-7 Índice Merck: 14,07107 |
|
Resultados test especificaciones
Appearance | Gray to Dark purple to Black powder to crystal |
Total Metals Impurities | max. 10.0 ppm |
Hole Mobility (mu FET) | min. 0.35 cm2/Vs (bare Si/SiO2 substrate) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...