4-Fenilpirimidina ≥96.0% (by GC, titration analysis)
Proveedor: TCI
|
Atención
|
P2525-1G
P2525-5G
TCIAP2525-1GEA
47
EUR
TCIAP2525-1G
TCIAP2525-5G
4-Fenilpirimidina ≥96.0% (by GC, titration analysis)
4-Fenilpirimidina
Formula:
C₁₀H₈N₂ MW: 156,19 g/mol Pto. Ebullición: 144°C |
Número MDL:
MFCD00006111 Núm. CAS: 3438-48-0 EINECS: 222-345-8 |
Resultados test especificaciones
Appearance | White to Yellow to Orange powder to crystal |
Purity (GC) | min. 96.0 % |
Purity (Nonaqueous Titration) | min. 96.0 % |
Melting point | 60.0 to 64.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...