Propionyl-L-carnitine hydrochloride ≥98.0% (by titrimetric analysis)

Proveedor: TCI
Atención

P2569-100MG P2569-1G
TCIAP2569-100MGEA 23 EUR
TCIAP2569-100MG TCIAP2569-1G
Propionyl-L-carnitine hydrochloride ≥98.0% (by titrimetric analysis)
Propionyl-L-carnitine hydrochloride
Formula: C₁₀H₁₉NO₄·HCl
Temperatura de almacenaje: Congelador
Número MDL: MFCD00274137
Núm. CAS: 119793-66-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=69bda2e7e4f0e483124ccf7dbae9a1d08db543b0&vid=69bda2e7e4f0e483124ccf7dbae9a1d08db543b0&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %
Purity (Argentmetric Titration) min. 98.0 %
Specific rotation [a]20/D -24.0 to -28.0 deg (C=1, H2O)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR