2-Fenil-9,10-antraquinona ≥98.0% (por GC)

Proveedor: TCI

P2588-1G P2588-5G
TCIAP2588-1GEA 55 EUR
TCIAP2588-1G TCIAP2588-5G
2-Fenil-9,10-antraquinona ≥98.0% (por GC)
2-Fenil-9,10-antraquinona
Formula: C₂₀H₁₂O₂
Número MDL: MFCD00559262
Núm. CAS: 6485-97-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3602b31da129947fdf1b586f201fb1a3e3b803a3&vid=3602b31da129947fdf1b586f201fb1a3e3b803a3&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Yellow to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 160.0 to 164.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR