N'-Hydroxypyridine-4-carboximidamide ≥98.0% (by HPLC, titration analysis)

Proveedor: TCI
Atención

Sinónimos: N-Hydroxyisonicotinimidamide, 4-Pyridylamidoxime, Pyridine-4-amidoxime, N'-Hydroxy-4-pyridinecarboximidamide

P2591-1G P2591-5G
TCIAP2591-1GEA 22 EUR
TCIAP2591-1G TCIAP2591-5G
N'-Hydroxypyridine-4-carboximidamide ≥98.0% (by HPLC, titration analysis)
N'-Hydroxypyridine-4-carboximidamide
Formula: C₆H₇N₃O
MW: 137,142 g/mol
Número MDL: MFCD00125873
Núm. CAS: 1594-57-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1a8960b70298cfcef0ee03f5746596b858bb20a7&vid=1a8960b70298cfcef0ee03f5746596b858bb20a7&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 203.0 to 207.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR