Phenylpiracetam ≥98.0% (by HPLC, total nitrogen)
Proveedor: TCI
P2604-1G
P2604-5G
TCIAP2604-1GEA
88
EUR
TCIAP2604-1G
TCIAP2604-5G
Phenylpiracetam ≥98.0% (by HPLC, total nitrogen)
Phenylpiracetam
Formula:
C₁₂H₁₄N₂O₂ |
Número MDL:
MFCD01456750 Núm. CAS: 77472-70-9 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Melting point | 130.0 to 134.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...