1,4-Phenylene bis(1,3-dioxo-1,3-dihydroisobenzofuran-5-carboxylate) ≥95.0% (by titrimetric analysis)

Proveedor: TCI

Sinónimos: Bis(1,3-dioxo-1,3-dihydroisobenzofuran-5-carboxylic acid) 1,4-Phenylene ester, TAHQ

P2625-25G P2625-5G
TCIAP2625-25GEA 496 EUR
TCIAP2625-25G TCIAP2625-5G
1,4-Phenylene bis(1,3-dioxo-1,3-dihydroisobenzofuran-5-carboxylate) ≥95.0% (by titrimetric analysis)
1,4-Phenylene bis(1,3-dioxo-1,3-dihydroisobenzofuran-5-carboxylate)
Formula: C₂₄H₁₀O₁₀
MW: 458,33 g/mol
Número MDL: MFCD00516832
Núm. CAS: 2770-49-2

Order Now

Resultados test especificaciones

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 93.0 area%
Purity (Morpholine Method) min. 95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR