Potassium Butyltrifluoroborate ≥98.0% (by titrimetric analysis)

Proveedor: TCI
Peligro

P2626-1G P2626-5G
TCIAP2626-1GEA 29 EUR
TCIAP2626-1G TCIAP2626-5G
Potassium Butyltrifluoroborate ≥98.0% (by titrimetric analysis)
Potassium Butyltrifluoroborate
Formula: C₄H₉BF₃K
Número MDL: MFCD08276805
Núm. CAS: 444343-55-9
UN: 3261
ADR: 8,II

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (Gravimetric) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR