Potassium ethyltrifluoroborate ≥97.0% (W)

Proveedor: TCI
Atención

P2628-1G P2628-5G
TCIAP2628-1GEA 72 EUR
TCIAP2628-1G TCIAP2628-5G
Potassium ethyltrifluoroborate ≥97.0% (W)
Potassium ethyltrifluoroborate
Formula: C₂H₅BF₃K
Número MDL: MFCD04112713
Núm. CAS: 882871-21-8

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (Gravimetric) min. 97.0 %

ESPECIFICACIONES

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR