Potasio (3-bromofenil)trifluoroborato ≥98.0% (by HPLC, titration analysis)
Proveedor: TCI
|
Atención
|
Sinónimos:
potassium (3-bromophenyl)trifluoroborate, (3-Bromophenyl)trifluoroboric acid potassium salt
P2646-1G
P2646-5G
TCIAP2646-1GEA
38
EUR
TCIAP2646-1G
TCIAP2646-5G
Potasio (3-bromofenil)trifluoroborato ≥98.0% (by HPLC, titration analysis)
Potasio (3-bromofenil)trifluoroborato
Formula:
C₆H₄BBrF₃K MW: 262.91 g/mol |
Número MDL:
MFCD02093977 Núm. CAS: 374564-34-8 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 186.0 to 191.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...