2-Fenil-1,3-ditiano ≥98.0% (por GC)

Proveedor: TCI

P2660-5G P2660-25G
TCIAP2660-5GEA 52 EUR
TCIAP2660-5G TCIAP2660-25G
2-Fenil-1,3-ditiano ≥98.0% (por GC)
2-Fenil-1,3-ditiano
Formula: C₁₀H₁₂S₂
MW: 196,34 g/mol
Número MDL: MFCD00006656
Núm. CAS: 5425-44-5
EINECS: 226-568-1

Order Now

Resultados test especificaciones

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 71.0 to 75.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR