4-(Pyridin-3-yl)pyrimidin-2-amine ≥98.0% (by GC, titration analysis)
Proveedor: TCI
|
Atención
|
P2666-1G
P2666-5G
TCIAP2666-1GEA
50
EUR
TCIAP2666-1G
TCIAP2666-5G
4-(Pyridin-3-yl)pyrimidin-2-amine ≥98.0% (by GC, titration analysis)
4-(Pyridin-3-yl)pyrimidin-2-amine
Formula:
C₉H₈N₄ |
Número MDL:
MFCD01317832 Núm. CAS: 66521-66-2 |
Resultados test especificaciones
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 189.0 to 193.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...