Pyridoxamine dihydrochloride ≥98.0% (by HPLC, total nitrogen)

Proveedor: TCI
Atención

P2688-1G P2688-25G P2688-5G
TCIAP2688-1GEA 27 EUR
TCIAP2688-1G TCIAP2688-25G TCIAP2688-5G
Pyridoxamine dihydrochloride ≥98.0% (by HPLC, total nitrogen)
Pyridoxamine dihydrochloride
Formula: C₈H₁₂N₂O₂·2HCl
Pto. de fusión: 227°C
Temperatura de almacenaje: Congelador
Número MDL: MFCD00012808
Núm. CAS: 524-36-7
EINECS: 208-357-6
Índice Merck: 14,07980

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Solubility in Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR