1,2,4-Triphenylbenzene ≥97.0% (por GC)
Proveedor: TCI
Sinónimos:
4'-Phenyl-1,1':2',1''-terphenyl
P2693-1G
P2693-200MG
TCIAP2693-1GEA
158
EUR
TCIAP2693-1G
TCIAP2693-200MG
1,2,4-Triphenylbenzene ≥97.0% (por GC)
1,2,4-Triphenylbenzene
Formula:
C₂₄H₁₈ MW: 306,41 g/mol |
Número MDL:
MFCD32062934 Núm. CAS: 1165-53-3 |
Resultados test especificaciones
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (GC) | min. 97.0 % |
Melting point | 117.0 to 121.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...