trans-4-(4-n-Pentylcyclohexyl)phenyl-trans,trans-4'-n-propylbicyclohexyl-4-carboxylate ≥98.0% (por GC)

Proveedor: TCI

P2696-1G P2696-5G
TCIAP2696-1GEA 48 EUR
TCIAP2696-1G TCIAP2696-5G
trans-4-(4-n-Pentylcyclohexyl)phenyl-trans,trans-4'-n-propylbicyclohexyl-4-carboxylate ≥98.0% (por GC)
trans-4-(4-n-Pentylcyclohexyl)phenyl-trans,trans-4'-n-propylbicyclohexyl-4-carboxylate
Formula: C₃₃H₅₂O₂
Número MDL: MFCD11053459
Núm. CAS: 131790-57-3

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Mesomorphic range 211.0 to 237.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR