Polyethylene glycol dimethacrylate n=∼9 estabilizado

Proveedor: TCI
Atención

Sinónimos: PEGDMA, Polyethylene Glycol Dimethacrylate average Mn 550, PEG dimethacrylate, Polyethylene Glycol Dimethacrylate

Estabilización: Stabilised with 4-methoxyphenol (MEHQ)

P2709-25G P2709-500G
TCIAP2709-25GEA 22 EUR
TCIAP2709-25G TCIAP2709-500G
Polyethylene glycol dimethacrylate n=∼9 estabilizado
Polyethylene glycol dimethacrylate
Formula: (C₂H₄O)nC₈H₁₀O₃
Número MDL: MFCD00014932
Núm. CAS: 25852-47-5

Order Now

Resultados test especificaciones

Appearance Colorless to Light yellow clear liquid
The average value of n 8.5 to 9.5

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR