Quinidine sulphate dihydrate ≥98.0% (by titrimetric analysis)

Proveedor: TCI
Peligro

Sinónimos: (S)-(6-methoxyquinolin-4-yl)((1S,2R,4S,5R)-5-vinylquinuclidin-2-yl)methanol sulphate hydrate(2:1:2)

Q0010-25G Q0010-5G
TCIAQ0010-25GEA 120 EUR
TCIAQ0010-25G TCIAQ0010-5G
Quinidine sulphate dihydrate ≥98.0% (by titrimetric analysis)
Quinidine sulphate dihydrate
Formula: C₄₀H₄₈N₄O₄.H₂O₄S·2H₂O
Número MDL: MFCD00149346
Núm. CAS: 6591-63-5
UN: 2811
ADR: 6.1,III
Índice Merck: 14,08060

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 80.0 area%
Purity (Neutralization titration) min. 98.0 % (calcd.on anh.substance)
Specific rotation [a]20/D +268 to +280 deg (C=2, 0.1mol/L HCl)
Water 4.0 to 5.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR