Quinuclidina ≥96.0% (by GC, titration analysis)

Proveedor: TCI
Peligro

Sinónimos: 1-Azabicyclo[2.2.2]octane, ABCO

Q0062-200MG Q0062-1G Q0062-5G
TCIAQ0062-200MGEA 27 EUR
TCIAQ0062-200MG TCIAQ0062-1G TCIAQ0062-5G
Quinuclidina ≥96.0% (by GC, titration analysis)
Quinuclidina
Formula: C₇H₁₃N
MW: 111,19 g/mol
Número MDL: MFCD00006690
Núm. CAS: 100-76-5
EINECS: 202-887-1
UN: 2811
ADR: 6.1,II
Índice Merck: 14,08081
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f788fb0934c3db157492f70b34c022eab299f7ff&vid=f788fb0934c3db157492f70b34c022eab299f7ff&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Light yellow powder to crystal
Purity (GC) min. 96.0 %
Purity (Neutralization titration) min. 96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR