8-Quinolineboronic acid (contains varying amounts of Anhydride)
Proveedor: TCI
|
Atención
|
Sinónimos:
Quinolin-8-ylboronic acid
Q0086-200MG
Q0086-1G
TCIAQ0086-200MGEA
32
EUR
TCIAQ0086-200MG
TCIAQ0086-1G
8-Quinolineboronic acid (contains varying amounts of Anhydride)
8-Quinolineboronic acid
Formula:
C₉H₈BNO₂ |
Número MDL:
MFCD01114698 Núm. CAS: 86-58-8 Índice Merck: 14,08069 |
|
Resultados test especificaciones
Appearance | White to Light yellow powder to crystal |
Purity (Neutralization titration) | 97.0 to 112.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...