6-Quinolinecarboxaldehyde ≥98.0% (by GC, titration analysis)
Proveedor: TCI
|
Atención
|
Q0097-1G
Q0097-5G
TCIAQ0097-1GEA
63
EUR
TCIAQ0097-1G
TCIAQ0097-5G
6-Quinolinecarboxaldehyde ≥98.0% (by GC, titration analysis)
6-Quinolinecarboxaldehyde
Formula:
C₁₀H₇NO |
Número MDL:
MFCD00805836 Núm. CAS: 4113-04-6 |
|
Resultados test especificaciones
Appearance | White to Yellow to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 74.0 to 78.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...