Sodium ricinolate ≥93.0% (by titrimetric analysis)
Proveedor: TCI
Sinónimos:
Ricinolic acid Sodium Salt, Sodium (R,Z)-12-hydroxyoctadec-9-enoate, Ricinoleic acid Sodium Salt
R0030-25G
R0030-500G
TCIAR0030-25GEA
31
EUR
TCIAR0030-25G
TCIAR0030-500G
Sodium ricinolate ≥93.0% (by titrimetric analysis)
Sodium ricinolate
Formula:
C₁₈H₃₄O₃ MW: 320.45 g/mol |
Número MDL:
MFCD00050760 Núm. CAS: 5323-95-5 EINECS: 226-191-2 Índice Merck: 14,08211 |
Resultados test especificaciones
Appearance | White to Orange to Green powder to crystal |
Purity (Nonaqueous Titration) | min. 93.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...