β-D-Ribofuranose-1,2,3,5-tetraacetate ≥98.0% (por GC)

Proveedor: TCI

Sinónimos: Tetra-O-acetyl-β-D-ribofuranose, (2S,3R,4R,5R)-5-(Acetoxymethyl)tetrahydrofuran-2,3,4-triyl triacetate

R0066-25G R0066-5G
TCIAR0066-25GEA 74 EUR
TCIAR0066-25G TCIAR0066-5G
β-D-Ribofuranose-1,2,3,5-tetraacetate ≥98.0% (por GC)
beta-D-Ribofuranose-1,2,3,5-tetraacetate
Formula: C₁₃H₁₈O₉
MW: 318,28 g/mol
Número MDL: MFCD00005358
Núm. CAS: 13035-61-5
EINECS: 235-898-5

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 82.0 to 85.0 deg-C
Specific rotation [a]20/D -13.0 to -16.0 deg (C=5, MeOH)
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR