Risperidone ≥98.0% (by HPLC, titration analysis)

Proveedor: TCI
Peligro

Sinónimos: 3-[2-[4-(6-Fluoro-1,2-benzisoxazol-3-yl)-1-piperidinyl]ethyl]-6,7,8,9-tetrahydro-2-methyl-4H-pyrido[1,2-a]pyrimidin-4-one, 3-(2-(4-(6-Fluorobenzo[d]isoxazol-3-yl)piperidin-1-yl)ethyl)-2-methyl-6,7,8,9-tetrahydro-4H-pyrido[1,2-a]pyrimidin-4-one

R0087-50MG R0087-500MG
TCIAR0087-50MGEA 16 EUR
TCIAR0087-50MG TCIAR0087-500MG
Risperidone ≥98.0% (by HPLC, titration analysis)
Risperidone
Formula: C₂₃H₂₇FN₄O₂
MW: 410,49 g/mol
Número MDL: MFCD00274576
Núm. CAS: 106266-06-2
UN: 2811
ADR: 6.1,III
Índice Merck: 14,08233

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 170.0 to 174.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR