Rivastigmine-L-tartrate ≥98.0% (by HPLC, titration analysis)

Proveedor: TCI
Peligro

Sinónimos: Rivastigmine tartrate, (S)-3-(1-(Dimethylamino)ethyl)phenyl ethyl(methyl)carbamate (2R,3R)-2,3-dihydroxysuccinate, N-Ethyl-N-methylcarbamic Acid 3-[(S)-1-(Dimethylamino)ethyl]phenyl Ester L-Tartrate, 3-[(S)-1-(Dimethylamino)ethyl]phenyl N-Ethyl-N-methylcarbamate L-Tartrate

R0093-1G R0093-5G
TCIAR0093-1GEA 94 EUR
TCIAR0093-1G TCIAR0093-5G
Rivastigmine-L-tartrate ≥98.0% (by HPLC, titration analysis)
Rivastigmine-L-tartrate
Formula: C₁₄H₂₂N₂O₂·C₄H₆O₆
MW: 400,43 g/mol
Número MDL: MFCD03700731
Núm. CAS: 129101-54-8
UN: 2811
ADR: 6.1,III

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 124.0 to 128.0 deg-C
Specific rotation [a]20/D +4.0 to +7.0 deg (C=5, methanol)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR