2-[[[4-(3-Methoxypropoxy)-3-methylpyridine-2-yl ]methyl]thio]-1H-benzimidazole ≥98.0% (by HPLC, titration analysis)
Proveedor: TCI
Sinónimos:
Rabeprazole sulphide, 2-[[4-(3-Methoxypropoxy)-3-methyl-2-pyridyl]methylthio]benzimidazole
R0098-200MG
R0098-1G
TCIAR0098-200MGEA
44
EUR
TCIAR0098-200MG
TCIAR0098-1G
2-[[[4-(3-Methoxypropoxy)-3-methylpyridine-2-yl ]methyl]thio]-1H-benzimidazole ≥98.0% (by HPLC, titration analysis)
2-[[[4-(3-Methoxypropoxy)-3-methylpyridine-2-yl ]methyl]thio]-1H-benzimidazole
Formula:
C₁₈H₂₁N₃O₂S MW: 343,4432 g/mol |
Número MDL:
MFCD08063845 Núm. CAS: 117977-21-6 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 114.0 to 118.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...