Ru-MACHO®-BH
Proveedor: TCI
|
Peligro
|
Sinónimos:
Carbonylhydrido(tetrahydroborato)[bis(2-diphenylphosphinoethyl)amino]ruthenium(II)
R0137-200MG
R0137-1G
TCIAR0137-200MGEA
35
EUR
TCIAR0137-200MG
TCIAR0137-1G
Ru-MACHO®-BH
Ru-MACHO®-BH
Formula:
C₂₉H₃₄BNOP₂Ru MW: 586,42 g/mol |
Número MDL:
MFCD20922901 Núm. CAS: 1295649-41-0 |
Resultados test especificaciones
Appearance | White to Yellow to Orange powder to crystal |
Content (Ruthenium) | 15 to 17 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...