(S)-RUCY®-XylBINAP

Proveedor: TCI
Peligro

Sinónimos: RuCl[(S)-daipena][(S)-xylbinap]

R0138-200MG R0138-1G
TCIAR0138-200MGEA 39 EUR
TCIAR0138-200MG TCIAR0138-1G
(S)-RUCY®-XylBINAP
(S)-RUCY®-XylBINAP
Formula: C₇₁H₇₃ClN₂O₂P₂Ru
MW: 1184,84 g/mol
Número MDL: MFCD20922903
Núm. CAS: 1312713-89-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=73486037ce9c83094e94a3fc63958f90fa7c6b7c&vid=36350ef0413cfaa45104777103955810ca7c7566&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance Yellow to Brown to Dark green powder to crystal
Content (Ruthenium) 8 to 9 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR