[RuCl(p-cymene)((R)-segphos®)]Cl

Proveedor: TCI

Sinónimos: Chloro[(R)-(+)-5,5'-bis(diphenylphosphino)-4,4'-bi-1,3-benzodioxole](p-cymene)ruthenium(II) chloride [RuCl(p-cymene)((R)-segphos®)]Cl, Chloro[(R)-(+)-5,5'-bis(diphenylphosphino)-4,4'-bi-1,3-benzodioxole](p-cymene)ruthenium(II) Chloride

R0154-200MG R0154-1G
TCIAR0154-200MGEA 36 EUR
TCIAR0154-200MG TCIAR0154-1G
[RuCl(p-cymene)((R)-segphos®)]Cl
[RuCl(p-cymene)((R)-segphos®)]Cl
Formula: C₄₈H₄₂Cl₂O₄P₂Ru
MW: 916,78 g/mol
Número MDL: MFCD09753017
Núm. CAS: 944451-28-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6203c8c0e47bac7f934236dbd38da5140400c68d&vid=6203c8c0e47bac7f934236dbd38da5140400c68d&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance Orange to Brown to Dark red powder to crystal
IR to pass test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR