[RuCl(p-cymene)((S)-segphos®)]Cl

Proveedor: TCI

Sinónimos: Chloro[(S)-(-)-5,5'-bis(diphenylphosphino)-4,4'-bi-1,3-benzodioxole](p-cymene)ruthenium(II) Chloride, Chloro[(S)-(-)-5,5'-bis(diphenylphosphino)-4,4'-bi-1,3-benzodioxole](p-cymene)ruthenium(II) chloride [RuCl(p-cymene)((S)-segphos®)]Cl

R0155-200MG R0155-1G
TCIAR0155-200MGEA 33 EUR
TCIAR0155-200MG TCIAR0155-1G
[RuCl(p-cymene)((S)-segphos®)]Cl
[RuCl(p-cymene)((S)-segphos®)]Cl
Formula: C₄₈H₄₂Cl₂O₄P₂Ru
Número MDL: MFCD09753017
Núm. CAS: 944451-29-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f98932c8df12177777dd342acad7327a0532b56f&vid=f98932c8df12177777dd342acad7327a0532b56f&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance Light yellow to Brown to Dark green powder to crystal
IR to pass test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR