[RuCl(p-cymene)((S)-dm-segphos®)]Cl
Proveedor: TCI
Sinónimos:
Chloro{(S)-(-)-5,5'-bis[di(3,5-xylyl)phosphino]-4,4'-bi-1,3-benzodioxole}(p-cymene)ruthenium(II) chloride [RuCl(p-cymene)((S)-dm-segphos®)]Cl, Chloro[(S)-(-)-5,5'-bis[di(3,5-xylyl)phosphino]-4,4'-bi-1,3-benzodioxole](p-cymene)ruthenium(II) Chloride
R0157-200MG
R0157-1G
TCIAR0157-200MGEA
37
EUR
TCIAR0157-200MG
TCIAR0157-1G
[RuCl(p-cymene)((S)-dm-segphos®)]Cl
[RuCl(p-cymene)((S)-dm-segphos®)]Cl
Formula:
C₅₆H₅₈Cl₂O₄P₂Ru MW: 1028,99 g/mol |
Número MDL:
MFCD09753018 Núm. CAS: 944451-31-4 |
Resultados test especificaciones
Appearance | Light yellow to Brown powder to crystal |
IR | to pass test |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...