Chloro{(S)-(+)-5,5'-bis[di(3,5-di-t-butyl-4-methoxyphenyl)phosphino]-4,4'-bi-1,3-benzodioxole}(p-cymene)ruthenium(II) chloride
Proveedor: TCI
Sinónimos:
[RuCl(p-cymene)((S)-dtbm-segphos®)]Cl, Chloro{(S)-(+)-5,5'-bis[di(3,5-di-t-butyl-4-methoxyphenyl)phosphino]-4,4'-bi-1,3-benzodioxole}(p-cymene)ruthenium(II) chloride [RuCl(p-cymene)((S)-dtbm-segphos®)]Cl
R0159-200MG
R0159-1G
TCIAR0159-200MGEA
33
EUR
TCIAR0159-200MG
TCIAR0159-1G
Chloro{(S)-(+)-5,5'-bis[di(3,5-di-t-butyl-4-methoxyphenyl)phosphino]-4,4'-bi-1,3-benzodioxole}(p-cymene)ruthenium(II) chloride
Chloro{(S)-(+)-5,5'-bis[di(3,5-di-t-butyl-4-methoxyphenyl)phosphino]-4,4'-bi-1,3-benzodioxole}(p-cymene)ruthenium(II) chloride
Formula:
C₈₄H₁₁₄Cl₂O₈P₂Ru MW: 1485,75 g/mol |
Número MDL:
MFCD10567030 Núm. CAS: 944451-33-6 |
Resultados test especificaciones
Appearance | Light yellow to Amber to Dark green powder to crystal |
IR | to pass test |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...