Rhodium(II) octanoate dimer

Proveedor: TCI

R0161-200MG R0161-1G
TCIAR0161-200MGEA 215 EUR
TCIAR0161-200MG TCIAR0161-1G
Rhodium(II) octanoate dimer
Rhodium(II) octanoate dimer
Formula: C₃₂H₆₀O₈Rh₂
Número MDL: MFCD00064724
Núm. CAS: 73482-96-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a4a407c2a5e883ae6b8a920eebcfcd7f87423beb&vid=a4a407c2a5e883ae6b8a920eebcfcd7f87423beb&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance Green to Dark green powder to crystal
Elemental analysis (Carbon) 47.00 to 52.00 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR