Rhodamine 640 Perchlorate ≥90.0% (by HPLC)
Proveedor: TCI
|
Atención
|
R0188-200MG
R0188-1G
TCIAR0188-200MGEA
62
EUR
TCIAR0188-200MG
TCIAR0188-1G
Rhodamine 640 Perchlorate ≥90.0% (by HPLC)
Rhodamine 640 Perchlorate
Formula:
C₃₂H₃₁ClN₂O₇ MW: 591,06 g/mol |
Número MDL:
MFCD00799440 Núm. CAS: 72102-91-1 EINECS: 276-345-8 UN: 1479 ADR: 5.1,III |
Resultados test especificaciones
Appearance | Amber to Brown to Dark green powder to crystal |
Purity (HPLC) | min. 90.0 area% |
Lambda max. | 560.0 to 575.0 nm (MeOH) |
Absorbance (E1%1cm) | min. 1800 (MeOH, 560 to 575nm) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...