Monosodium succinate ≥98.0% (by titrimetric analysis)

Proveedor: TCI
Atención

Sinónimos: Sodium Hydrogen Succinate, Succinic Acid Monosodium Salt, Sodium Bisuccinate

S0106-25G S0106-500G
TCIAS0106-25GEA 16 EUR
TCIAS0106-25G TCIAS0106-500G
Monosodium succinate ≥98.0% (by titrimetric analysis)
Monosodium succinate
Formula: C₄H₆O₄
MW: 140.07 g/mol
Número MDL: MFCD00050993
Núm. CAS: 2922-54-5
EINECS: 220-871-2

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Solubility in Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR