Dibenzyl sebacate ≥98.0% (por GC)
Proveedor: TCI
Sinónimos:
Decanedioic Acid Dibenzyl Ester, Sebacic Acid Dibenzyl Ester
S0147-25G
S0147-5G
TCIAS0147-25GEA
122
EUR
TCIAS0147-25G
TCIAS0147-5G
Dibenzyl sebacate ≥98.0% (por GC)
Dibenzyl sebacate
Formula:
C₂₄H₃₀O₄ MW: 382.50 g/mol |
Número MDL:
MFCD00039640 Núm. CAS: 140-24-9 EINECS: 205-404-2 |
|
Resultados test especificaciones
Appearance | White or Colorless to Light orange to Yellow powder to lump to clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...