p-Sexiphenyl

Proveedor: TCI

S0220-1G S0220-100MG
TCIAS0220-1GEA 762 EUR
TCIAS0220-1G TCIAS0220-100MG
p-Sexiphenyl
p-Sexiphenyl
Formula: C₃₆H₂₆
Número MDL: MFCD00039559
Núm. CAS: 4499-83-6
EINECS: 224-799-2

Order Now

Resultados test especificaciones

Appearance White to Orange to Green powder to crystaline
TOF-MS to pass test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR