β-Sitosterol acetate 40,00% (por GC) (contains Campesterol Acetate)

Proveedor: TCI

S0221-1G S0221-5G
TCIAS0221-1GEA 55 EUR
TCIAS0221-1G TCIAS0221-5G
β-Sitosterol acetate 40,00% (por GC) (contains Campesterol Acetate)
beta-Sitosterol acetate
Formula: C₃₁H₅₂O₂
MW: 456,76 g/mol
Número MDL: MFCD00083497
Núm. CAS: 915-05-9
EINECS: 213-019-6

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to lump
Purity (GC) min. 40.0 %
Melting point 131.0 to 136.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR