Magnesio estearato

Proveedor: TCI
Atención

Sinónimos: Stearic acid Magnesium(II) salt, Magnesium(II) Stearate, Stearic acid magnesium salt

S0238-25G S0238-250G
TCIAS0238-25GEA 16 EUR
TCIAS0238-25G TCIAS0238-250G
Magnesio estearato
Magnesio estearato
Formula: C₃₆H₇₀MgO₄
Número MDL: MFCD00036391
Núm. CAS: 557-04-0
EINECS: 209-150-3
Índice Merck: 14,05690
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cd7b328272ebf36e3aef3676b34324ee02fecf35&vid=cd7b328272ebf36e3aef3676b34324ee02fecf35&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Content of Magnesium 4.00 to 5.00 %
Palmitic acid (composition of fatty acid) max. 40.0 %
Stearic acid (composition of fatty acid) min. 60.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR