trans-Stilbene oxide ≥98.0% (by HPLC)

Proveedor: TCI

S0262-25G S0262-5G
TCIAS0262-25GEA 265 EUR
TCIAS0262-25G TCIAS0262-5G
trans-Stilbene oxide ≥98.0% (by HPLC)
trans-Stilbene oxide
Formula: C₁₄H₁₂O
Número MDL: MFCD00064311
Núm. CAS: 1439-07-2
EINECS: 215-877-7

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Melting point 67.0 to 73.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR