4-tert-Butylphenyl salicylate ≥98.0% (por GC)
Proveedor: TCI
|
Atención
|
Sinónimos:
Salicylic Acid 4-tert-Butylphenyl Ester
S0274-25G
S0274-500G
TCIAS0274-25GEA
19
EUR
TCIAS0274-25G
TCIAS0274-500G
4-tert-Butylphenyl salicylate ≥98.0% (por GC)
4-tert-Butylphenyl salicylate
Formula:
C₁₇H₁₈O₃ MW: 270,33 g/mol |
Número MDL:
MFCD00045759 Núm. CAS: 87-18-3 EINECS: 201-728-3 Índice Merck: 14,01586 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 63.0 to 66.0 deg-C |
Solubility in hot Methanol | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...