1-(3-sulphopropyl)pyridinium hydroxide inner salt ≥98.0% (by HPLC, total nitrogen)

Proveedor: TCI
Atención

S0391-25G S0391-250G
TCIAS0391-25GEA 21 EUR
TCIAS0391-25G TCIAS0391-250G
1-(3-sulphopropyl)pyridinium hydroxide inner salt ≥98.0% (by HPLC, total nitrogen)
1-(3-sulphopropyl)pyridinium hydroxide inner salt
Formula: C₈H₁₁NO₃S
Número MDL: MFCD00064468
Núm. CAS: 15471-17-7
EINECS: 239-491-3

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR