Methyl-3-sulpholene-3-carboxylate ≥98.0% (by HPLC)

Proveedor: TCI

Sinónimos: 2,5-Dihydro-3-methoxycarbonylthiophene 1,1-Dioxide, 3-sulpholene-3-carboxylic acid Methyl Ester, Methyl 2,5-dihydrothiophene-3-carboxylate 1,1-dioxide, 3-Methoxycarbonyl-3-sulpholene

S0476-1G S0476-5G
TCIAS0476-1GEA 65 EUR
TCIAS0476-1G TCIAS0476-5G
Methyl-3-sulpholene-3-carboxylate ≥98.0% (by HPLC)
Methyl-3-sulpholene-3-carboxylate
Formula: C₆H₈O₄S
MW: 176,19 g/mol
Número MDL: MFCD00042754
Núm. CAS: 67488-50-0
EINECS: 266-700-5

Order Now

Resultados test especificaciones

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Melting point 63.0 to 67.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR